Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026488.4 | NP_001021659.1 | 5 - 6 | 1285 | 69 | |
| NM_001026489.7 | NP_001021660.2 | 18 - 19 | 4251 | 69 | ||
| NM_001026490.6 | NP_001021661.2 | 18 - 19 | 3961 | 69 | ||
| NM_001306400.4 | NP_001293329.1 | 18 - 19 | 3965 | 69 | ||
| NM_001306401.3 | NP_001293330.1 | 13 - 14 | 3388 | 69 | ||
| NM_001306402.3 | NP_001293331.1 | 13 - 14 | 3100 | 69 | ||
| NM_001306403.3 | NP_001293332.1 | 12 - 13 | 3202 | 69 | ||
| NM_001306404.3 | NP_001293333.1 | 12 - 13 | 2914 | 69 | ||
| NM_001306405.3 | NP_001293334.1 | 9 - 10 | 2923 | 69 | ||
| NM_001306406.3 | NP_001293335.1 | 9 - 10 | 2635 | 69 | ||
| NM_001306407.3 | NP_001293336.1 | 7 - 8 | 2677 | 69 | ||
| NM_001306408.3 | NP_001293337.1 | 7 - 8 | 2389 | 69 | ||
| NM_001306529.4 | NP_001293458.1 | 8 - 9 | 3006 | 69 | ||
| NM_001356800.4 | NP_001343732.1 | 15 | 3804 | 69 | ||
| NM_058537.5 | NP_490938.3 | 18 - 19 | 4245 | 69 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map