Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026488.4 | NP_001021659.1 | 6 - 7 | 1807 | 63 | |
| NM_001026489.7 | NP_001021660.2 | 19 - 20 | 4773 | 63 | ||
| NM_001026490.6 | NP_001021661.2 | 19 - 20 | 4483 | 63 | ||
| NM_001306400.4 | NP_001293329.1 | 19 - 20 | 4487 | 63 | ||
| NM_001306401.3 | NP_001293330.1 | 14 - 15 | 3910 | 63 | ||
| NM_001306402.3 | NP_001293331.1 | 14 - 15 | 3622 | 63 | ||
| NM_001306403.3 | NP_001293332.1 | 13 - 14 | 3724 | 63 | ||
| NM_001306404.3 | NP_001293333.1 | 13 - 14 | 3436 | 63 | ||
| NM_001306405.3 | NP_001293334.1 | 10 - 11 | 3445 | 63 | ||
| NM_001306406.3 | NP_001293335.1 | 10 - 11 | 3157 | 63 | ||
| NM_001306407.3 | NP_001293336.1 | 8 - 9 | 3199 | 63 | ||
| NM_001306408.3 | NP_001293337.1 | 8 - 9 | 2911 | 63 | ||
| NM_001306529.4 | NP_001293458.1 | 9 - 10 | 3528 | 63 | ||
| NM_001356800.4 | NP_001343732.1 | 16 | 4326 | 63 | ||
| NM_058537.5 | NP_490938.3 | 19 - 20 | 4767 | 63 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map