Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 1 - 2 | 159 | 73 | |
| NM_001026490.6 | NP_001021661.2 | 1 - 2 | 163 | 73 | ||
| NM_001306400.4 | NP_001293329.1 | 1 - 2 | 161 | 73 | ||
| NM_058537.5 | NP_490938.3 | 1 - 2 | 159 | 73 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map