Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 6 - 7 | 995 | 96 | |
| NM_001026490.6 | NP_001021661.2 | 6 - 7 | 993 | 96 | ||
| NM_001306400.4 | NP_001293329.1 | 6 - 7 | 997 | 96 | ||
| NM_001306401.3 | NP_001293330.1 | 1 - 2 | 132 | 96 | ||
| NM_001306402.3 | NP_001293331.1 | 1 - 2 | 132 | 96 | ||
| NM_001356800.4 | NP_001343732.1 | 3 | 548 | 96 | ||
| NM_058537.5 | NP_490938.3 | 6 - 7 | 989 | 96 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map