Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 9 - 10 | 1316 | 69 | |
| NM_001026490.6 | NP_001021661.2 | 9 - 10 | 1314 | 69 | ||
| NM_001306400.4 | NP_001293329.1 | 9 - 10 | 1318 | 69 | ||
| NM_001306401.3 | NP_001293330.1 | 4 - 5 | 453 | 69 | ||
| NM_001306402.3 | NP_001293331.1 | 4 - 5 | 453 | 69 | ||
| NM_001306403.3 | NP_001293332.1 | 3 - 4 | 267 | 69 | ||
| NM_001306404.3 | NP_001293333.1 | 3 - 4 | 267 | 69 | ||
| NM_001356800.4 | NP_001343732.1 | 6 | 869 | 69 | ||
| NM_058537.5 | NP_490938.3 | 9 - 10 | 1310 | 69 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map