Assay Details
| Gene Symbol: cars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306359.3 | NP_001293288.1 | 3 - 4 | 650 | 71 | |
| NM_058612.3 | NP_491013.1 | 3 - 4 | 1115 | 71 | ||
| NM_058613.6 | NP_491014.1 | 3 - 4 | 1112 | 71 | ||
| Gene Symbol: Y23H5A.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131365.1 | 1 - 2 | 196 | 71 | ||
More Information
Set Membership: |
|

cars-1
Genomic Map