Assay Details
| Gene Symbol: cars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306359.3 | NP_001293288.1 | 6 - 7 | 2192 | 79 | |
| NM_058612.3 | NP_491013.1 | 6 - 7 | 2657 | 79 | ||
| NM_058613.6 | NP_491014.1 | 6 - 7 | 2654 | 79 | ||
More Information
Set Membership: |
|

cars-1
Genomic Map