Assay Details
| Gene Symbol: arx-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381571.2 | NP_001367661.1 | 8 - 9 | 1516 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arx-7| Gene Symbol: arx-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381571.2 | NP_001367661.1 | 8 - 9 | 1516 | 73 | |
Set Membership: |
|