Assay Details
| Gene Symbol: neto-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058758.5 | NP_491159.3 | 3 - 4 | 301 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

neto-1| Gene Symbol: neto-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058758.5 | NP_491159.3 | 3 - 4 | 301 | 88 | |
Set Membership: |
|