Assay Details
| Gene Symbol: atic-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380345.2 | NP_001367679.1 | 1 - 2 | 61 | 103 | |
| NM_171912.7 | NP_741452.2 | 1 - 2 | 59 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380345.2, NM_171912.7 transcripts.
Set Membership: |
|

atic-1
Genomic Map