Assay Details
| Gene Symbol: atic-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027930.5 | NP_001023101.2 | 3 - 4 | 335 | 121 | |
| NM_001380345.2 | NP_001367679.1 | 7 - 8 | 727 | 121 | ||
| NM_171912.7 | NP_741452.2 | 7 - 8 | 725 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380345.2 transcripts.
Set Membership: |
|

atic-1
Genomic Map