Assay Details
| Gene Symbol: ubc-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060363.6 | NP_492764.2 | 4 - 5 | 1018 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ubc-25| Gene Symbol: ubc-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060363.6 | NP_492764.2 | 4 - 5 | 1018 | 62 | |
Set Membership: |
|