Assay Details
| Gene Symbol: rtel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060368.6 | NP_492769.1 | 1 - 2 | 406 | 93 | |
| Gene Symbol: F29C6.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381203.1 | NP_001367064.1 | 1 - 2 | 398 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381203.1 transcripts.
Set Membership: |
|

rtel-1
Genomic Map