Assay Details
| Gene Symbol: rtel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060368.6 | NP_492769.1 | 9 - 10 | 2327 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rtel-1| Gene Symbol: rtel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060368.6 | NP_492769.1 | 9 - 10 | 2327 | 92 | |
Set Membership: |
|