Assay Details
| Gene Symbol: trh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171427.4 | NP_741499.1 | 3 - 4 | 279 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trh-1| Gene Symbol: trh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171427.4 | NP_741499.1 | 3 - 4 | 279 | 111 | |
Set Membership: |
|