Assay Details
| Gene Symbol: ttn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001359765.3 | NP_001346723.1 | 43399 | 86 | ||
| NM_001359766.3 | NP_001346716.1 | 43444 | 86 | |||
| NM_001359767.3 | NP_001346722.1 | 44677 | 86 | |||
| NM_001359768.3 | NP_001346715.1 | 44722 | 86 | |||
| NM_001373034.1 | NP_001360072.1 | 44815 | 86 | |||
| NM_001380654.1 | NP_001368672.1 | 22 - 23 | 22687 | 86 | ||
| NM_001380655.1 | NP_001368671.1 | 23 - 24 | 22732 | 86 | ||
| NM_001380656.1 | NP_001367914.1 | 43399 | 86 | |||
| NM_001380657.1 | NP_001367912.1 | 43537 | 86 | |||
| NM_001380658.2 | NP_001367913.1 | 44677 | 86 | |||
| NM_001380659.1 | NP_001368674.1 | 22 - 23 | 22687 | 86 | ||
| NM_001380660.1 | NP_001368673.1 | 23 - 24 | 22732 | 86 | ||
More Information
Set Membership: |
|

ttn-1
Genomic Map