Assay Details
| Gene Symbol: osm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171545.9 | NP_741633.1 | 4 - 5 | 397 | 146 | |
| GenBank mRNA | AJ000259.1 | - | 4 - 5 | 357 | 146 | |
More Information
Set Membership: |
|

osm-6
Genomic Map