Assay Details
| Gene Symbol: osm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171545.9 | NP_741633.1 | 6 - 7 | 660 | 67 | |
| GenBank mRNA | AJ000259.1 | - | 6 - 7 | 620 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

osm-6| Gene Symbol: osm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171545.9 | NP_741633.1 | 6 - 7 | 660 | 67 | |
| GenBank mRNA | AJ000259.1 | - | 6 - 7 | 620 | 67 | |
Set Membership: |
|