Assay Details
| Gene Symbol: myrf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270166.4 | NP_001257095.1 | 2 - 3 | 300 | 126 | |
| NM_001381079.1 | NP_001367016.1 | 2 - 3 | 209 | 126 | ||
| NM_001381080.1 | NP_001367259.1 | 1 - 2 | 105 | 126 | ||
| NM_001383635.1 | 2 - 3 | 229 | 126 | |||
| NM_077308.7 | NP_509709.2 | 2 - 3 | 120 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383635.1 transcripts.
Set Membership: |
|

myrf-2
Genomic Map