Assay Details
| Gene Symbol: myrf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270166.4 | NP_001257095.1 | 4 - 5 | 743 | 62 | |
| NM_001381079.1 | NP_001367016.1 | 4 - 5 | 652 | 62 | ||
| NM_001381080.1 | NP_001367259.1 | 3 - 4 | 548 | 62 | ||
| NM_001383635.1 | 4 - 5 | 672 | 62 | |||
| NM_077308.7 | NP_509709.2 | 4 - 5 | 563 | 62 | ||
More Information
Set Membership: |
|

myrf-2
Genomic Map