Assay Details
| Gene Symbol: myrf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270166.4 | NP_001257095.1 | 6 - 7 | 1233 | 108 | |
| NM_001381079.1 | NP_001367016.1 | 6 - 7 | 1142 | 108 | ||
| NM_001381080.1 | NP_001367259.1 | 5 - 6 | 1038 | 108 | ||
| NM_001383635.1 | 6 - 7 | 1162 | 108 | |||
| NM_077308.7 | NP_509709.2 | 6 - 7 | 1053 | 108 | ||
More Information
Set Membership: |
|

myrf-2
Genomic Map