Assay Details
| Gene Symbol: myrf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270166.4 | NP_001257095.1 | 8 - 9 | 2367 | 70 | |
| NM_001381079.1 | NP_001367016.1 | 8 - 9 | 2276 | 70 | ||
| NM_001381080.1 | NP_001367259.1 | 7 - 8 | 2172 | 70 | ||
| NM_001383635.1 | 8 - 9 | 2296 | 70 | |||
| NM_077308.7 | NP_509709.2 | 8 - 9 | 2187 | 70 | ||
More Information
Set Membership: |
|

myrf-2
Genomic Map