Assay Details
| Gene Symbol: plk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026002.5 | NP_001021173.1 | 2 - 3 | 296 | 66 | |
| GenBank mRNA | AF080581.1 | - | 1 - 2 | 272 | 66 | |
More Information
Set Membership: |
|

plk-1
Genomic Map