Assay Details
| Gene Symbol: atic-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027930.5 | NP_001023101.2 | 8 - 9 | 883 | 142 | |
| NM_001380345.2 | NP_001367679.1 | 13 - 14 | 1326 | 142 | ||
| NM_171912.7 | NP_741452.2 | 12 - 13 | 1273 | 142 | ||
More Information
Set Membership: |
|

atic-1
Genomic Map