Assay Details
| Gene Symbol: gyg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076207.8 | NP_508608.1 | 4 - 5 | 611 | 76 | |
| NM_076208.7 | NP_508609.1 | 4 - 5 | 611 | 76 | ||
| NM_171944.8 | NP_741749.1 | 5 - 6 | 756 | 76 | ||
More Information
Set Membership: |
|

gyg-1
Genomic Map