Assay Details
| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372639.1 | NP_001359523.1 | 1 - 2 | 404 | 106 | |
| NM_001382955.1 | 1 - 2 | 411 | 106 | |||
| NM_181898.4 | NP_871627.1 | 1 - 2 | 404 | 106 | ||
| NM_181899.7 | NP_871628.1 | 1 - 2 | 411 | 106 | ||
| Gene Symbol: R107.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066600.5 | NP_499001.1 | 1 - 2 | 404 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_066600.5 transcripts.
Set Membership: |
|

ikke-1
Genomic Map