Assay Details
| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181898.4 | NP_871627.1 | 4 - 5 | 2245 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ikke-1| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181898.4 | NP_871627.1 | 4 - 5 | 2245 | 95 | |
Set Membership: |
|