Assay Details
| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181898.4 | NP_871627.1 | 5 - 6 | 2323 | 75 | |
| NM_181899.7 | NP_871628.1 | 5 - 6 | 2318 | 75 | ||
More Information
Set Membership: |
|

ikke-1
Genomic Map