Assay Details
| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382955.1 | 4 - 5 | 2254 | 83 | ||
| NM_181899.7 | NP_871628.1 | 4 - 5 | 2254 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ikke-1| Gene Symbol: ikke-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382955.1 | 4 - 5 | 2254 | 83 | ||
| NM_181899.7 | NP_871628.1 | 4 - 5 | 2254 | 83 | ||
Set Membership: |
|