Assay Details
| Gene Symbol: mtm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067352.6 | NP_499753.1 | 3 - 4 | 326 | 61 | |
| NM_181951.6 | NP_871680.1 | 3 - 4 | 641 | 61 | ||
| GenBank mRNA | AY313177.1 | - | 3 - 4 | 306 | 61 | |
| AY313179.1 | - | 3 - 4 | 624 | 61 | ||
More Information
Set Membership: |
|

mtm-6
Genomic Map