Assay Details
| Gene Symbol: cpna-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026713.3 | NP_001021884.1 | 10 - 11 | 3640 | 151 | |
| NM_063222.4 | NP_495623.2 | 10 - 11 | 3640 | 151 | ||
| NM_063224.6 | NP_495625.3 | 10 - 11 | 3640 | 151 | ||
More Information
Set Membership: |
|

cpna-2
Genomic Map