Assay Details
| Gene Symbol: cat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267080.3 | NP_001254009.1 | 3 - 4 | 785 | 85 | |
| NM_001306554.2 | NP_001293483.1 | 3 - 4 | 716 | 85 | ||
| NM_001383774.1 | 2 - 3 | 1422 | 85 | |||
More Information
Set Membership: |
|

cat-2
Genomic Map