Assay Details
| Gene Symbol: mag-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060624.8 | NP_493025.1 | 2 - 3 | 170 | 94 | |
| GenBank mRNA | KF051002.1 | - | 1 - 2 | 62 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mag-1| Gene Symbol: mag-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060624.8 | NP_493025.1 | 2 - 3 | 170 | 94 | |
| GenBank mRNA | KF051002.1 | - | 1 - 2 | 62 | 94 | |
Set Membership: |
|