Assay Details
| Gene Symbol: ksr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026347.5 | NP_001021518.1 | 6 - 7 | 1381 | 128 | |
| NM_001026348.4 | NP_001021519.1 | 6 - 7 | 1261 | 128 | ||
| NM_001136307.4 | NP_001129779.1 | 4 - 5 | 1089 | 128 | ||
| GenBank mRNA | AY077614.1 | - | 6 - 7 | 1381 | 128 | |
| AY077615.1 | - | 6 - 7 | 1261 | 128 | ||
More Information
Set Membership: |
|

ksr-2
Genomic Map