Assay Details
| Gene Symbol: pad-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060851.7 | NP_493252.3 | 6 - 7 | 1415 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pad-1| Gene Symbol: pad-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060851.7 | NP_493252.3 | 6 - 7 | 1415 | 65 | |
Set Membership: |
|