Assay Details
| Gene Symbol: gpi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026667.7 | NP_001021838.1 | 2 - 3 | 367 | 65 | |
| NM_001383747.1 | 2 - 3 | 204 | 65 | |||
| Gene Symbol: ubc-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383748.1 | 2 - 3 | 221 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383748.1 transcripts.
Set Membership: |
|

gpi-1
Genomic Map