Assay Details
| Gene Symbol: smd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383756.1 | 3 - 4 | 1132 | 59 | ||
| GenBank mRNA | Y12499.1 | - | 1145 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smd-1| Gene Symbol: smd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383756.1 | 3 - 4 | 1132 | 59 | ||
| GenBank mRNA | Y12499.1 | - | 1145 | 59 | ||
Set Membership: |
|