Assay Details
| Gene Symbol: gopc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026706.4 | NP_001021877.2 | 4 - 5 | 703 | 79 | |
| NM_001026707.5 | NP_001021878.2 | 4 - 5 | 784 | 79 | ||
| NM_001129097.5 | NP_001122569.1 | 4 - 5 | 566 | 79 | ||
More Information
Set Membership: |
|

gopc-1
Genomic Map