Assay Details
| Gene Symbol: eya-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025884.6 | NP_001021055.1 | 7 - 8 | 1170 | 69 | |
| NM_001381258.2 | NP_001367120.1 | 7 - 8 | 1076 | 69 | ||
More Information
Set Membership: |
|

eya-1
Genomic Map