Assay Details
| Gene Symbol: mrps-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061179.7 | NP_493580.1 | 5 - 6 | 1152 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrps-27| Gene Symbol: mrps-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061179.7 | NP_493580.1 | 5 - 6 | 1152 | 83 | |
Set Membership: |
|