Assay Details
| Gene Symbol: cat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267080.3 | NP_001254009.1 | 7 - 8 | 1539 | 83 | |
| NM_001306554.2 | NP_001293483.1 | 7 - 8 | 1470 | 83 | ||
| NM_001383774.1 | 6 - 7 | 2176 | 83 | |||
More Information
Set Membership: |
|

cat-2
Genomic Map