Assay Details
| Gene Symbol: mtq-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061808.4 | NP_494209.2 | 1 - 2 | 109 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtq-2| Gene Symbol: mtq-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061808.4 | NP_494209.2 | 1 - 2 | 109 | 86 | |
Set Membership: |
|