Assay Details
| Gene Symbol: srpa-72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061965.6 | NP_494366.2 | 4 - 5 | 752 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srpa-72| Gene Symbol: srpa-72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061965.6 | NP_494366.2 | 4 - 5 | 752 | 85 | |
Set Membership: |
|