Assay Details
| Gene Symbol: ntl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047261.5 | NP_001040726.1 | 3 - 4 | 399 | 66 | |
| NM_001383818.1 | 5 - 6 | 795 | 66 | |||
| NM_062371.5 | NP_494772.1 | 6 - 7 | 715 | 66 | ||
| NM_062372.6 | NP_494773.1 | 4 - 5 | 461 | 66 | ||
More Information
Set Membership: |
|

ntl-2
Genomic Map