Assay Details
| Gene Symbol: tbck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377792.1 | NP_001364726.1 | 9 - 10 | 1720 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbck-1| Gene Symbol: tbck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377792.1 | NP_001364726.1 | 9 - 10 | 1720 | 124 | |
Set Membership: |
|