Assay Details
| Gene Symbol: fut-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062705.10 | NP_495106.1 | 6 - 7 | 1291 | 79 | |
| GenBank mRNA | AJ745072.1 | - | 2 - 3 | 445 | 79 | |
More Information
Set Membership: |
|

fut-3
Genomic Map