Assay Details
| Gene Symbol: fut-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062705.10 | NP_495106.1 | 7 - 8 | 1506 | 100 | |
| GenBank mRNA | AJ745072.1 | - | 3 - 4 | 660 | 100 | |
More Information
Set Membership: |
|

fut-3
Genomic Map