Assay Details
| Gene Symbol: fut-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062705.10 | NP_495106.1 | 9 - 10 | 1733 | 92 | |
| GenBank mRNA | AJ745072.1 | - | 5 - 6 | 887 | 92 | |
More Information
Set Membership: |
|

fut-3
Genomic Map