Assay Details
| Gene Symbol: ift-43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062706.3 | NP_495107.1 | 2 - 3 | 158 | 75 | |
| Gene Symbol: fut-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062705.10 | NP_495106.1 | 1 - 2 | 248 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_062705.10 transcripts.
Set Membership: |
|

ift-43
Genomic Map