Assay Details
| Gene Symbol: tbc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062755.9 | NP_495156.1 | 4 - 5 | 821 | 125 | |
| Gene Symbol: ZK1248.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383850.1 | 7 - 8 | 1421 | 125 | ||
More Information
Set Membership: |
|

tbc-2
Genomic Map